Learn More
Waters Corp Cartridge, C18, 1.6 μm, 2.1 x 5 mm
Catalog No. 502391210 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These VanGuard FIT products are designed to improve analytical column lifetime by removing particulate and chemical contamination from the mobile-phase stream without compromising chromatographic performance.
  • Minimal dispersion, allowing column protection without sacrificing results.
  • No additional tubing, which reduces the risk of leaks.
  • Consistent connections lead to consistent results.
  • Easy to install with just a quarter turn to tighten.
  • Restores analytical column performance and backpressure to like new with a quick and simple change.
  • Returns column backpressure to baseline each time the cartridge is replaced.
  • Features 1.6 μm particle size.
  • Dimensions are 2.1 x 5 mm.
  • Utilizes C18 chemistry.
Catalog No. 50-239-1210 Supplier Waters Corp Supplier No. 186009517
These VanGuard FIT products are designed to improve analytical column lifetime by removing particulate and chemical contamination from the mobile-phase stream without compromising chromatographic performance.
  • Minimal dispersion, allowing column protection without sacrificing results.
  • No additional tubing, which reduces the risk of leaks.
  • Consistent connections lead to consistent results.
  • Easy to install with just a quarter turn to tighten.
  • Restores analytical column performance and backpressure to like new with a quick and simple change.
  • Returns column backpressure to baseline each time the cartridge is replaced.
  • Features 1.6 μm particle size.
  • Dimensions are 2.1 x 5 mm.
  • Utilizes C18 chemistry.