Learn More
Waters Corp Column, C18+, 2.7 μm, 4.6 mm × 150 mm
Catalog No. 502392376 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These general purpose, high efficiency C18 columns are built on a solid-core particle. They are designed to deliver excellent peak shape for basic analytes at low pH. MaxPeak High Performance Surface hardware technology significantly reduces unwanted analyte/surface interactions, improving peak shape and signal intensity. The VanGuard Fit column design protects the column from contaminants and extends its lifetime.
  • General purpose, high efficiency C18 columns based on a solid-core particle.
  • Designed to deliver excellent peak shape for basic analytes at low pH.
  • Utilizes MaxPeak High Performance Surface hardware technology to significantly reduce unwanted analyte/surface interactions, improving peak shape and signal intensity.
  • VanGuard Fit column design protects the column from contaminants and increases column lifetime.
Catalog No. 50-239-2376 Supplier Waters Corp Supplier No. 186010470
These general purpose, high efficiency C18 columns are built on a solid-core particle. They are designed to deliver excellent peak shape for basic analytes at low pH. MaxPeak High Performance Surface hardware technology significantly reduces unwanted analyte/surface interactions, improving peak shape and signal intensity. The VanGuard Fit column design protects the column from contaminants and extends its lifetime.
  • General purpose, high efficiency C18 columns based on a solid-core particle.
  • Designed to deliver excellent peak shape for basic analytes at low pH.
  • Utilizes MaxPeak High Performance Surface hardware technology to significantly reduce unwanted analyte/surface interactions, improving peak shape and signal intensity.
  • VanGuard Fit column design protects the column from contaminants and increases column lifetime.