Learn More
Waters Corp Column, Premier C18, VanGuard FIT, 2.7 μm
Catalog No. 502390795 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The CORTECS Premier C18 2.7 μm 4.6 X 150 mm Column is a high-efficiency, solid-core column with MaxPeak High Performance Surface technology and VanGuard Fit design.
  • General purpose, high-efficiency column technology.
  • Solid-core particle for optimal performance.
  • Balanced retention for acids, bases, and neutrals across various pH levels.
  • MaxPeak High Performance Surface hardware for reduced analyte/surface interactions.
  • Improved peak shape and signal intensity.
  • VanGuard Fit design for contamination protection.
  • Extended column lifetime.
  • C18 chemistry.
  • Reversed phase separation mode.
  • Endcapped for enhanced stability.
  • Parker-style endfitting type.
  • 4.6 mm inner diameter.
  • 150 mm length.
  • 6.6 % carbon load.
Catalog No. 50-239-0795 Supplier Waters Corp Supplier No. 186010453
The CORTECS Premier C18 2.7 μm 4.6 X 150 mm Column is a high-efficiency, solid-core column with MaxPeak High Performance Surface technology and VanGuard Fit design.
  • General purpose, high-efficiency column technology.
  • Solid-core particle for optimal performance.
  • Balanced retention for acids, bases, and neutrals across various pH levels.
  • MaxPeak High Performance Surface hardware for reduced analyte/surface interactions.
  • Improved peak shape and signal intensity.
  • VanGuard Fit design for contamination protection.
  • Extended column lifetime.
  • C18 chemistry.
  • Reversed phase separation mode.
  • Endcapped for enhanced stability.
  • Parker-style endfitting type.
  • 4.6 mm inner diameter.
  • 150 mm length.
  • 6.6 % carbon load.