Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Column, CORTECS C18, 2.7 μm, 4.6 × 100 mm

Catalog No. 502392303 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

The CORTECS Premier C18 VanGuard FIT Column, featuring inert stainless steel hardware, incorporates MaxPeak High Performance Surfaces (HPS) and C18 chemistry to deliver enhanced chromatographic performance. MaxPeak HPS technology is engineered to minimize analyte/surface interactions, improving analyte recovery, sensitivity, and reproducibility, especially for metal-sensitive compounds like lipids, organic acids, and peptides. The integrated VanGuard FIT technology further protects the column, maintaining separation efficiency and extending its operational lifetime.

  • Reduces column conditioning and passivation times.
  • Improves sensitivity and peak shapes.
  • Enhances recovery for various analytes.
  • Maximizes efficiency from solid-core technology.
  • Lowers column backpressure.
  • Particle size: 2.7 μm.
  • Dimensions: 4.6 mm × 100 mm.
  • Chemistry: C18.

Catalog No. 50-239-2303 Supplier Waters Corp Supplier No. 186010452
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The CORTECS Premier C18 VanGuard FIT Column, featuring inert stainless steel hardware, incorporates MaxPeak High Performance Surfaces (HPS) and C18 chemistry to deliver enhanced chromatographic performance. MaxPeak HPS technology is engineered to minimize analyte/surface interactions, improving analyte recovery, sensitivity, and reproducibility, especially for metal-sensitive compounds like lipids, organic acids, and peptides. The integrated VanGuard FIT technology further protects the column, maintaining separation efficiency and extending its operational lifetime.

  • Reduces column conditioning and passivation times.
  • Improves sensitivity and peak shapes.
  • Enhances recovery for various analytes.
  • Maximizes efficiency from solid-core technology.
  • Lowers column backpressure.
  • Particle size: 2.7 μm.
  • Dimensions: 4.6 mm × 100 mm.
  • Chemistry: C18.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.