Learn More
Waters Corp Column, CORTECS Premier C18+, 2.7 μm, 2.1 mm X 50 mm
Catalog No. 502390872 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The CORTECS Premier C18+ 2.7μm 2.1 X 50 mm Columns are general purpose, high efficiency C18 columns based on a solid-core particle. They are designed to deliver excellent peak shape for basic analytes at low pH. These columns utilize MaxPeak High Performance Surface hardware technology, which significantly reduces unwanted analyte/surface interactions that can lead to poor peak shape and losses in signal intensity. The VanGuard Fit column design protects the column from contaminants and increases column lifetime.
  • Delivers excellent peak shape for basic analytes at low pH.
  • MaxPeak High Performance Surface hardware technology reduces unwanted analyte/surface interactions.
  • Reduces poor peak shape and losses in signal intensity.
  • VanGuard Fit column design protects the column from contaminants.
  • Increases column lifetime.
  • Documents are easily accessible online to reduce waste.
Catalog No. 50-239-0872 Supplier Waters Corp Supplier No. 186010458
The CORTECS Premier C18+ 2.7μm 2.1 X 50 mm Columns are general purpose, high efficiency C18 columns based on a solid-core particle. They are designed to deliver excellent peak shape for basic analytes at low pH. These columns utilize MaxPeak High Performance Surface hardware technology, which significantly reduces unwanted analyte/surface interactions that can lead to poor peak shape and losses in signal intensity. The VanGuard Fit column design protects the column from contaminants and increases column lifetime.
  • Delivers excellent peak shape for basic analytes at low pH.
  • MaxPeak High Performance Surface hardware technology reduces unwanted analyte/surface interactions.
  • Reduces poor peak shape and losses in signal intensity.
  • VanGuard Fit column design protects the column from contaminants.
  • Increases column lifetime.
  • Documents are easily accessible online to reduce waste.