Learn More
Waters Corp C18 Column, 1.6 μm, 2.1 x 150 mm, VanGuard FIT
Supplier:  Waters Corp 186009516
Encompass_Preferred
The CORTECS Premier C18 VanGuard FIT Column from Waters is a general purpose, high-efficiency column based on a solid-core particle. It is designed to offer balanced retention of acids, bases, and neutrals at low and mid-range pH. This column utilizes MaxPeak High Performance Surface hardware technology, which significantly reduces unwanted analyte/surface interactions that can lead to poor peak shape and losses in signal intensity.
  • General purpose, high-efficiency columns
  • Based on a solid-core particle
  • Offers balanced retention of acids, bases and neutrals at low and mid-range pH
  • Uses MaxPeak High Performance Surface hardware technology which significantly reduces unwanted analyte/surface interactions that can lead to poor peak shape and losses in signal intensity
  • VanGuard Fit column design protects the column from contaminants and increases column lifetime
Catalog No. 50-239-0614