Learn More
Waters Corp CORTECS Column, C18, 1.6 μm, 2.1 mm X 50 mm
Supplier:  Waters Corp 186009514
Encompass_Preferred
These high-efficiency columns are built on a solid-core particle, providing balanced retention for acids, bases, and neutrals across low to mid-range pH conditions. MaxPeak High Performance Surface hardware technology minimizes unwanted analyte interactions, improving peak shape and signal intensity. The VanGuard Fit design safeguards the column from contaminants, extending its operational lifespan.
  • High-efficiency columns based on a solid-core particle
  • Offers balanced retention of acids, bases, and neutrals at low and mid-range pH
  • Uses MaxPeak High Performance Surface hardware technology to reduce unwanted analyte/surface interactions, improving peak shape and signal intensity
  • VanGuard Fit column design protects the column from contaminants
  • Increases column lifetime
Catalog No. 50-239-0636