Learn More
Waters Corp CORTECS Premier FIT Column, 5 μm, C18, 2.1 x 5 mm
Catalog No. NC3947414 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Waters™ VanGuard™ FIT Products are designed to improve analytical column lifetime by removing particulate and chemical contamination from the mobile-phase stream without compromising chromatographic performance. They offer minimal dispersion, no additional tubing, consistent connections, and are easy to install with a quarter-turn to tighten. These products can restore analytical column performance and backpressure to like new with a quick and simple change of the VanGuard FIT Cartridge.
  • Minimal dispersion
  • No additional tubing
  • Consistent connections
  • Easy to install with a quarter-turn to tighten
  • Restores analytical column performance and backpressure
  • Quick and simple cartridge change
  • 2.1 x 5 mm cartridge dimensions
  • C18 chemistry
  • 5 μm particle size
  • Available in a 3-pack
Catalog No. NC3947414 Supplier Waters Corp Supplier No. 186010870
Waters™ VanGuard™ FIT Products are designed to improve analytical column lifetime by removing particulate and chemical contamination from the mobile-phase stream without compromising chromatographic performance. They offer minimal dispersion, no additional tubing, consistent connections, and are easy to install with a quarter-turn to tighten. These products can restore analytical column performance and backpressure to like new with a quick and simple change of the VanGuard FIT Cartridge.
  • Minimal dispersion
  • No additional tubing
  • Consistent connections
  • Easy to install with a quarter-turn to tighten
  • Restores analytical column performance and backpressure
  • Quick and simple cartridge change
  • 2.1 x 5 mm cartridge dimensions
  • C18 chemistry
  • 5 μm particle size
  • Available in a 3-pack