Learn More
Waters Corp CORTECS Premier T3 Column, 1.6 μm, 2.1 mm x 150 mm
Catalog No. NC2294351 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
CORTECS Premier T3 Columns are designed for high-performance separation of polar and non-polar analytes. Utilizing MaxPeak High Performance Surface technology and T3 bonding, these columns reduce unwanted interactions and offer high efficiency with lower backpressure for superior analytical results.
  • Employs T3 bonding technology
  • Particle morphology offers high efficiency and lower backpressure compared to fully porous particles of similar size
  • Offers ultra-low column MS bleed
  • Ensures long lifetimes
  • Delivers excellent peak shape
This item is not returnable. View return policy
Catalog No. NC2294351 Supplier Waters Corp Supplier No. 186010436
CORTECS Premier T3 Columns are designed for high-performance separation of polar and non-polar analytes. Utilizing MaxPeak High Performance Surface technology and T3 bonding, these columns reduce unwanted interactions and offer high efficiency with lower backpressure for superior analytical results.
  • Employs T3 bonding technology
  • Particle morphology offers high efficiency and lower backpressure compared to fully porous particles of similar size
  • Offers ultra-low column MS bleed
  • Ensures long lifetimes
  • Delivers excellent peak shape