Learn More
Waters Corp CORTECS Premier T3 Column, 1.6 μm, 2.1 mm, 100 mm
Supplier:  Waters Corp 186010435
Encompass_Preferred
CORTECS Premier Columns, including the T3 Column, utilize solid-core particle technology and MaxPeak High Performance Surfaces (HPS) for superior chromatographic performance. They deliver exceptional peak shapes, low backpressure, and reliable data for diverse LC-MS applications.
  • Solid-core particles enable higher resolution, faster throughput, and better separation.
  • MaxPeak HPS minimizes unwanted metal interactions, improving peak shape and data quality for metal-sensitive analytes.
  • Versatile for small molecule and biomolecule LC-MS applications across UHPLC, HPLC, and legacy LC platforms.
  • Offers scalable performance and consistent results across various particle sizes.
  • Low backpressure design permits high-speed separations.
  • Constructed with high-grade stainless steel or inert materials to minimize sample interactions.
Catalog No. NC2294340