Learn More
Waters Corp Waters CORTECS Premier T3 Column, 1.6 μm, 2.1 mm, 50 mm
Catalog No. NC2294338 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These CORTECS Premier T3 Columns offer excellent retention for polar and non-polar analytes with 100% aqueous mobile phase compatibility. Their particle morphology ensures high efficiency and lower backpressure. MaxPeak High Performance Surface hardware technology significantly reduces unwanted analyte/surface interactions, providing ultra-low column MS bleed, long lifetimes, and excellent peak shape.
  • Compatible with 100% aqueous mobile phases, providing excellent retention for polar and non-polar analytes.
  • Particle morphology offers high efficiency and lower backpressure compared to fully porous particles of similar size.
  • Provides ultra-low column MS bleed, long lifetimes, and excellent peak shape.
  • Utilizes MaxPeak High Performance Surface hardware technology to significantly reduce unwanted analyte/surface interactions, which can cause poor peak shape and signal intensity losses.
This item is not returnable. View return policy
Catalog No. NC2294338 Supplier Waters Corp Supplier No. 186010434
Add to Cart
Add to Cart
These CORTECS Premier T3 Columns offer excellent retention for polar and non-polar analytes with 100% aqueous mobile phase compatibility. Their particle morphology ensures high efficiency and lower backpressure. MaxPeak High Performance Surface hardware technology significantly reduces unwanted analyte/surface interactions, providing ultra-low column MS bleed, long lifetimes, and excellent peak shape.
  • Compatible with 100% aqueous mobile phases, providing excellent retention for polar and non-polar analytes.
  • Particle morphology offers high efficiency and lower backpressure compared to fully porous particles of similar size.
  • Provides ultra-low column MS bleed, long lifetimes, and excellent peak shape.
  • Utilizes MaxPeak High Performance Surface hardware technology to significantly reduce unwanted analyte/surface interactions, which can cause poor peak shape and signal intensity losses.