Learn More
Waters Corp Waters CORTECS Premier T3 Column, 2.7 μm, 4.6 mm, 50 mm
Supplier:  Waters Corp 186010479
Encompass_Preferred
This CORTECS Premier T3 Column is for HPLC and UHPLC applications. It features a 2.7 μm spherical silica particle with T3 bonding technology, providing excellent retention for polar and non-polar analytes. MaxPeak High Performance Surface hardware minimizes unwanted analyte interactions, ensuring optimal peak shape and signal intensity.
  • Compatible with 100% aqueous mobile phases.
  • Provides excellent retention for polar and non-polar analytes.
  • Offers high efficiency and lower backpressure compared to fully porous particles of similar size.
  • Provides ultra-low column MS bleed.
  • Long lifetimes.
  • Excellent peak shape.
  • Uses MaxPeak High Performance Surface hardware technology, which significantly reduces unwanted analyte/surface interactions that can lead to poor peak shape and losses in signal intensity.
Catalog No. NC2294376