Learn More
Waters Corp CORTECS Premier T3 Column, 2.7 μm, 2.1 x 150 mm, 120 Å
Catalog No. NC2294375 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
CORTECS Premier T3 Columns feature MaxPeak High Performance Surfaces, designed to enhance chromatographic separations by minimizing analyte/surface interactions. This technology improves analyte recovery, sensitivity, and reproducibility, and helps mitigate the loss of metal-sensitive analytes such as lipids, organic acids, acidic peptides, oligonucleotides, or other compounds containing phosphate or carboxylate functionalities.
  • Reduced column conditioning and passivation times.
  • Improved sensitivity and peak shapes.
  • Simpler mobile phases, without complex additives.
  • Time savings in method development.
  • Reduced risk and greater confidence in data and decision making.
This item is not returnable. View return policy
Catalog No. NC2294375 Supplier Waters Corp Supplier No. 186010474
CORTECS Premier T3 Columns feature MaxPeak High Performance Surfaces, designed to enhance chromatographic separations by minimizing analyte/surface interactions. This technology improves analyte recovery, sensitivity, and reproducibility, and helps mitigate the loss of metal-sensitive analytes such as lipids, organic acids, acidic peptides, oligonucleotides, or other compounds containing phosphate or carboxylate functionalities.
  • Reduced column conditioning and passivation times.
  • Improved sensitivity and peak shapes.
  • Simpler mobile phases, without complex additives.
  • Time savings in method development.
  • Reduced risk and greater confidence in data and decision making.