Learn More
Waters Corp Waters CORTECS Premier T3 HPLC Column, 2.7 μm, 4.6 mm ID, 100 mm L
Catalog No. NC2294377 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
CORTECS Premier T3 Columns are compatible with 100% aqueous mobile phases, offering excellent retention for polar and non-polar analytes. Their particle morphology ensures high efficiency and lower backpressure. These columns feature ultra-low MS bleed, long lifetimes, and excellent peak shape, utilizing MaxPeak High Performance Surface hardware technology to reduce unwanted analyte/surface interactions and improve signal intensity.
  • Compatible with 100% aqueous mobile phases.
  • Provides excellent retention for polar and non-polar analytes.
  • Offers high efficiency and lower backpressure.
  • Delivers ultra-low column MS bleed.
  • Ensures long lifetimes and excellent peak shape.
  • Utilizes MaxPeak High Performance Surface hardware technology to reduce unwanted analyte/surface interactions and improve signal intensity.
This item is not returnable. View return policy
Catalog No. NC2294377 Supplier Waters Corp Supplier No. 186010480
CORTECS Premier T3 Columns are compatible with 100% aqueous mobile phases, offering excellent retention for polar and non-polar analytes. Their particle morphology ensures high efficiency and lower backpressure. These columns feature ultra-low MS bleed, long lifetimes, and excellent peak shape, utilizing MaxPeak High Performance Surface hardware technology to reduce unwanted analyte/surface interactions and improve signal intensity.
  • Compatible with 100% aqueous mobile phases.
  • Provides excellent retention for polar and non-polar analytes.
  • Offers high efficiency and lower backpressure.
  • Delivers ultra-low column MS bleed.
  • Ensures long lifetimes and excellent peak shape.
  • Utilizes MaxPeak High Performance Surface hardware technology to reduce unwanted analyte/surface interactions and improve signal intensity.