Learn More
Waters Corp Waters Column, C18, 2.7 μm, 2.1 mm × 100 mm
Catalog No. NC2294374 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These columns are compatible with 100% aqueous mobile phases, and provide excellent retention for polar and non-polar analytes. Their particle morphology offers high efficiency and lower backpressure when compared to fully porous particles of similar particle size.
  • Provides ultra-low column MS bleed
  • Ensures long lifetimes
  • Offers excellent peak shape
  • Utilizes MaxPeak High Performance Surface hardware technology to reduce unwanted analyte/surface interactions
  • Prevents poor peak shape and losses in signal intensity with MaxPeak High Performance Surface hardware
This item is not returnable. View return policy
Catalog No. NC2294374 Supplier Waters Corp Supplier No. 186010473
These columns are compatible with 100% aqueous mobile phases, and provide excellent retention for polar and non-polar analytes. Their particle morphology offers high efficiency and lower backpressure when compared to fully porous particles of similar particle size.
  • Provides ultra-low column MS bleed
  • Ensures long lifetimes
  • Offers excellent peak shape
  • Utilizes MaxPeak High Performance Surface hardware technology to reduce unwanted analyte/surface interactions
  • Prevents poor peak shape and losses in signal intensity with MaxPeak High Performance Surface hardware