Learn More
Waters Corp CORTECS Premier T3 VanGuard FIT Column, 2.7 μm, 2.1 mm, 150 mm
Catalog No. 502392593 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The CORTECS Premier T3 2.7 μm 2.1 x 150 mm Columns are compatible with 100% aqueous mobile phases, offering excellent retention for polar and non-polar analytes. MaxPeak High Performance Surface hardware technology reduces unwanted analyte/surface interactions. The VanGuard Fit column design protects against contaminants and extends column lifetime.
  • Excellent retention for polar and non-polar analytes in aqueous mobile phases.
  • Reduces unwanted analyte/surface interactions.
  • Protects against contaminants and extends column lifetime.
  • High efficiency and lower backpressure.
  • Ultra-low MS bleed and long lifetimes.
Catalog No. 50-239-2593 Supplier Waters Corp Supplier No. 186010477
The CORTECS Premier T3 2.7 μm 2.1 x 150 mm Columns are compatible with 100% aqueous mobile phases, offering excellent retention for polar and non-polar analytes. MaxPeak High Performance Surface hardware technology reduces unwanted analyte/surface interactions. The VanGuard Fit column design protects against contaminants and extends column lifetime.
  • Excellent retention for polar and non-polar analytes in aqueous mobile phases.
  • Reduces unwanted analyte/surface interactions.
  • Protects against contaminants and extends column lifetime.
  • High efficiency and lower backpressure.
  • Ultra-low MS bleed and long lifetimes.