Learn More
Waters Corp CORTECS Premier T3 VanGuard FIT Column, 2.7 μm, 4.6 x 150 mm
Catalog No. 502391545 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
MaxPeak Premier Columns feature MaxPeak High Performance Surfaces (HPS), designed to improve chromatographic separations by minimizing unwanted analyte/surface interactions. By reducing these interactions and associated sample losses, MaxPeak HPS enhances analyte recovery, sensitivity, and reproducibility. These columns are beneficial for mitigating loss of metal-sensitive analytes such as lipids, organic acids, acidic peptides, oligonucleotides, nucleic acids, and plasmids. They are compatible with various particle technologies for separations in reversed-phase, HILIC, and SEC modes across diverse applications.
  • Reduced column conditioning and passivation times
  • Improved sensitivity and peak shapes
  • Improved recovery
  • Simpler mobile phases, eliminating the need for complex additives
  • Time savings in method development
  • Reduced risk and increased confidence in data and decision-making
Catalog No. 50-239-1545 Supplier Waters Corp Supplier No. 186010484
MaxPeak Premier Columns feature MaxPeak High Performance Surfaces (HPS), designed to improve chromatographic separations by minimizing unwanted analyte/surface interactions. By reducing these interactions and associated sample losses, MaxPeak HPS enhances analyte recovery, sensitivity, and reproducibility. These columns are beneficial for mitigating loss of metal-sensitive analytes such as lipids, organic acids, acidic peptides, oligonucleotides, nucleic acids, and plasmids. They are compatible with various particle technologies for separations in reversed-phase, HILIC, and SEC modes across diverse applications.
  • Reduced column conditioning and passivation times
  • Improved sensitivity and peak shapes
  • Improved recovery
  • Simpler mobile phases, eliminating the need for complex additives
  • Time savings in method development
  • Reduced risk and increased confidence in data and decision-making