Learn More
Waters Corp Column, C18, 1.6 μm, 2.1 mm × 100 mm
Catalog No. 502391630 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The CORTECS Premier T3 1.6 μm 2.1 × 100 mm Columns are compatible with 100% aqueous mobile phases, offer excellent retention for polar and non-polar analytes, and deliver ultra-low column MS bleed, long lifetimes, and excellent peak shape. MaxPeak High Performance Surface hardware reduces unwanted analyte/surface interactions. VanGuard Fit design protects from contaminants and extends column lifetime.
  • Compatible with 100% aqueous mobile phases; excellent retention for polar and non-polar analytes.
  • High efficiency, lower backpressure, ultra-low MS bleed, long lifetimes, and excellent peak shape.
  • MaxPeak High Performance Surface hardware technology reduces unwanted analyte/surface interactions.
  • VanGuard Fit column design protects from contaminants and increases column lifetime.
Catalog No. 50-239-1630 Supplier Waters Corp Supplier No. 186010438
The CORTECS Premier T3 1.6 μm 2.1 × 100 mm Columns are compatible with 100% aqueous mobile phases, offer excellent retention for polar and non-polar analytes, and deliver ultra-low column MS bleed, long lifetimes, and excellent peak shape. MaxPeak High Performance Surface hardware reduces unwanted analyte/surface interactions. VanGuard Fit design protects from contaminants and extends column lifetime.
  • Compatible with 100% aqueous mobile phases; excellent retention for polar and non-polar analytes.
  • High efficiency, lower backpressure, ultra-low MS bleed, long lifetimes, and excellent peak shape.
  • MaxPeak High Performance Surface hardware technology reduces unwanted analyte/surface interactions.
  • VanGuard Fit column design protects from contaminants and increases column lifetime.