Learn More
Waters Corp CORTECS Premier T3 VanGuard FIT Column, 2.7 μm, 2.1 mm x 50 mm, C18
Catalog No. 502392380 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These columns are compatible with 100% aqueous mobile phases, providing excellent retention for polar and non-polar analytes. They offer high efficiency and lower backpressure due to unique particle morphology. MaxPeak High Performance Surface hardware and VanGuard Fit design reduce unwanted analyte interactions, increasing lifetime.
  • Compatible with 100% aqueous mobile phases for polar and non-polar analyte retention.
  • Achieves high efficiency and lower backpressure with unique particle morphology.
  • Ensures ultra-low column MS bleed, long lifetimes, and excellent peak shape.
  • MaxPeak High Performance Surface technology reduces unwanted analyte/surface interactions.
  • VanGuard Fit design protects the column from contaminants and extends lifetime.
  • Easy to install with a quarter-turn tightening.
  • Restores analytical column performance by simple cartridge replacement.
Catalog No. 50-239-2380 Supplier Waters Corp Supplier No. 186010475
These columns are compatible with 100% aqueous mobile phases, providing excellent retention for polar and non-polar analytes. They offer high efficiency and lower backpressure due to unique particle morphology. MaxPeak High Performance Surface hardware and VanGuard Fit design reduce unwanted analyte interactions, increasing lifetime.
  • Compatible with 100% aqueous mobile phases for polar and non-polar analyte retention.
  • Achieves high efficiency and lower backpressure with unique particle morphology.
  • Ensures ultra-low column MS bleed, long lifetimes, and excellent peak shape.
  • MaxPeak High Performance Surface technology reduces unwanted analyte/surface interactions.
  • VanGuard Fit design protects the column from contaminants and extends lifetime.
  • Easy to install with a quarter-turn tightening.
  • Restores analytical column performance by simple cartridge replacement.