Learn More
Waters Corp CORTECS Premier T3 VanGuard FIT Column, 2.7 μm, 4.6 x 50 mm
Supplier:  Waters Corp 186010482
Encompass_Preferred
The CORTECS Premier T3 VanGuard FIT Column (2.7 μm, 4.6 x 50 mm) is designed to extend analytical column lifetime by efficiently removing particulate and chemical contamination from the mobile-phase stream without compromising chromatographic performance.
  • Offers minimal dispersion, ensuring results are not sacrificed, and features no additional tubing, which reduces the risk of leaks.
  • Provides consistent connections for consistent results and is easy to install with just a quarter-turn to tighten.
  • Maintains analytical column performance and can restore column performance and backpressure to like-new condition with a quick and simple change of the VanGuard FIT Cartridge.
Catalog No. 50-239-1539