Learn More
Waters Corp VanGuard FIT Cartridge, 1.6 μm, 2.1 X 5 mm, T3
Catalog No. 502391638 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The VanGuard FIT Column Protection is designed to extend the lifetime of analytical columns by filtering out particulate and chemical contaminants from the mobile-phase stream without compromising chromatographic performance. It protects the analytical column from degradation caused by sample impurities, such as proteins, sugars, and fats, thereby maintaining consistent analytical column performance and backpressure.
  • Minimal dispersion, ensuring column protection without sacrificing results.
  • No additional tubing, which reduces the risk of leaks.
  • Consistent connections, leading to consistent results.
  • Easy installation process requiring just a quarter turn to tighten.
  • Improved analytical column lifetime.
  • Ability to restore analytical column performance and backpressure to original levels by simply changing the cartridge.
Catalog No. 50-239-1638 Supplier Waters Corp Supplier No. 186010440
The VanGuard FIT Column Protection is designed to extend the lifetime of analytical columns by filtering out particulate and chemical contaminants from the mobile-phase stream without compromising chromatographic performance. It protects the analytical column from degradation caused by sample impurities, such as proteins, sugars, and fats, thereby maintaining consistent analytical column performance and backpressure.
  • Minimal dispersion, ensuring column protection without sacrificing results.
  • No additional tubing, which reduces the risk of leaks.
  • Consistent connections, leading to consistent results.
  • Easy installation process requiring just a quarter turn to tighten.
  • Improved analytical column lifetime.
  • Ability to restore analytical column performance and backpressure to original levels by simply changing the cartridge.