Learn More
Waters Corp CORTECS Premier FIT Column, 1.6 μm, T3, 2.1 x 150 mm
Catalog No. 502391999 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Waters™ VanGuard™ FIT Products are designed to extend the lifetime of analytical columns by removing particulate and chemical contamination from the mobile-phase stream without compromising chromatographic performance.
  • Minimal dispersion – protects the column without sacrificing results.
  • No additional tubing – reduces the risk of leaks.
  • Consistent connections – ensures consistent results.
  • Easy to install – requires just a quarter turn to tighten.
Catalog No. 50-239-1999 Supplier Waters Corp Supplier No. 186010439
Waters™ VanGuard™ FIT Products are designed to extend the lifetime of analytical columns by removing particulate and chemical contamination from the mobile-phase stream without compromising chromatographic performance.
  • Minimal dispersion – protects the column without sacrificing results.
  • No additional tubing – reduces the risk of leaks.
  • Consistent connections – ensures consistent results.
  • Easy to install – requires just a quarter turn to tighten.