Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp VanGuard FIT Cartridge, 2.7 μm, 2.1 × 5 mm

Catalog No. 502392504 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

Waters Corporation VanGuard FIT Cartridge, 2.7 μm, 2.1 × 5 mm

Catalog No. 50-239-2504 Supplier Waters Corp Supplier No. 186010478
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Waters Corporation VanGuard FIT Cartridge, 2.7 μm, 2.1 × 5 mm

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.