Learn More
Waters Corp CORTECS Shield RP18 Column, 90Å, 2.7 μm, 2.1 mm X 75 mm
Catalog No. 502390984 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
CORTECS Shield RP18 columns provide complementary selectivity to C8 and C18 stationary phases. The embedded carbamate group in the bonded phase ligand provides alternative selectivity, especially for phenolic compounds, compared to straight-chain alkyl columns. The solid-core particle morphology offers high efficiency and lower backpressures when compared to fully porous particles of a similar size.
  • Provides complementary selectivity to C8 and C18 stationary phases.
  • Embedded carbamate group in the bonded phase ligand provides alternative selectivity, especially for phenolic compounds.
  • Solid core particle morphology delivers high efficiency and reduced backpressures.
Catalog No. 50-239-0984 Supplier Waters Corp Supplier No. 186008663
CORTECS Shield RP18 columns provide complementary selectivity to C8 and C18 stationary phases. The embedded carbamate group in the bonded phase ligand provides alternative selectivity, especially for phenolic compounds, compared to straight-chain alkyl columns. The solid-core particle morphology offers high efficiency and lower backpressures when compared to fully porous particles of a similar size.
  • Provides complementary selectivity to C8 and C18 stationary phases.
  • Embedded carbamate group in the bonded phase ligand provides alternative selectivity, especially for phenolic compounds.
  • Solid core particle morphology delivers high efficiency and reduced backpressures.