Learn More
Waters Corp CORTECS Shield RP18 Column, C18, Reversed Phase, 2.7 μm, 4.6 mm × 150 mm
Catalog No. 502391005 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These columns offer complementary selectivity to C8 and C18 stationary phases with an embedded carbamate group for alternate selectivity, especially for phenolic compounds. Its solid core particle morphology delivers high efficiency and lower backpressures, providing balanced retention of acids, bases, and neutrals for excellent peak shape of basic analytes at low pH.
  • Provides complementary selectivity to C8 and C18 stationary phases.
  • Embedded carbamate group offers alternate selectivity for phenolic compounds.
  • Solid core particle morphology ensures high efficiency and lower backpressures.
  • Delivers high-efficiency results and balanced retention.
  • Achieves excellent peak shape for basic analytes at low pH.
  • Improves performance by lowering all three terms of the van Deemter equation.
  • Enables increased sample throughput by raising method flow rate.
Catalog No. 50-239-1005 Supplier Waters Corp Supplier No. 186008685
These columns offer complementary selectivity to C8 and C18 stationary phases with an embedded carbamate group for alternate selectivity, especially for phenolic compounds. Its solid core particle morphology delivers high efficiency and lower backpressures, providing balanced retention of acids, bases, and neutrals for excellent peak shape of basic analytes at low pH.
  • Provides complementary selectivity to C8 and C18 stationary phases.
  • Embedded carbamate group offers alternate selectivity for phenolic compounds.
  • Solid core particle morphology ensures high efficiency and lower backpressures.
  • Delivers high-efficiency results and balanced retention.
  • Achieves excellent peak shape for basic analytes at low pH.
  • Improves performance by lowering all three terms of the van Deemter equation.
  • Enables increased sample throughput by raising method flow rate.