Learn More
Waters Corp CORTECS Shield RP18 Column, 2.7 μm, 90Å, 3 mm x 30 mm
Supplier:  Waters Corp 176003917
Encompass_Preferred
The CORTECS Shield RP18 column offers complementary selectivity to C8 and C18 stationary phases, utilizing an embedded carbamate group in its bonded phase ligand for alternate selectivity, particularly with phenolic compounds. Its solid core particle morphology provides high efficiency and lower backpressures compared to fully porous particles of similar size. This column is designed to be all-purpose, delivering effective results and excellent peak shapes for basic analytes at low pH by balancing the retention of bases, acids, and neutrals.
  • Provides alternate selectivity for phenolic compounds compared to straight chain alkyl columns.
  • Offers high efficiency and lower backpressures.
  • Achieves excellent peak shapes for basic analytes at low pH.
  • Increases sample throughput with equivalent or superior peak capacities.
Catalog No. 50-239-1586