Learn More
Waters Corp CORTECS Shield RP18 Column, 90Å, 2.7 μm, 2.1 mm × 50 mm
Catalog No. 502391657 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These columns provide complementary selectivity to C8 and C18 stationary phases. The embedded carbamate group in the bonded phase ligand offers alternate selectivity, especially for phenolic compounds, compared to straight-chain alkyl columns. The solid core particle morphology ensures high efficiency and lower backpressures when compared to fully porous particles of similar particle size.
  • Provides complementary selectivity to C8 and C18 stationary phases.
  • Embedded carbamate group in bonded phase ligand offers alternate selectivity, particularly for phenolic compounds.
  • Solid core particle morphology contributes to high efficiency.
  • Ensures lower backpressures compared to fully porous particles of similar size.
Catalog No. 50-239-1657 Supplier Waters Corp Supplier No. 176003913
These columns provide complementary selectivity to C8 and C18 stationary phases. The embedded carbamate group in the bonded phase ligand offers alternate selectivity, especially for phenolic compounds, compared to straight-chain alkyl columns. The solid core particle morphology ensures high efficiency and lower backpressures when compared to fully porous particles of similar particle size.
  • Provides complementary selectivity to C8 and C18 stationary phases.
  • Embedded carbamate group in bonded phase ligand offers alternate selectivity, particularly for phenolic compounds.
  • Solid core particle morphology contributes to high efficiency.
  • Ensures lower backpressures compared to fully porous particles of similar size.