Learn More
Waters Corp Column, 90Å, 1.6 μm, 2.1 mm x 50 mm
Supplier:  Waters Corp 186008692
Encompass_Preferred
This Shield RP18 column offers complementary selectivity to C8 and C18 stationary phases. Its embedded carbamate group provides alternate selectivity, while solid core particle morphology ensures high efficiency and lower backpressures.
  • Complementary selectivity to C8 and C18 stationary phases.
  • Embedded carbamate group for alternate selectivity, especially for phenolic compounds.
  • Solid core particle morphology provides high efficiency and lower backpressures.
  • Balanced retention of acids, bases, and neutrals at low and mid-range pH.
  • Excellent peak shape for basic analytes even at low pH.
  • Solid-core particles decrease van Deemter equation components for improved performance.
  • Increased sample throughput by higher flow rates without compromising separation.
  • Operates efficiently between pH 2 to 8, up to 45 °C and 18,000 psi (1240 Bar).
Catalog No. 50-239-1899