Learn More
Waters Corp CORTECS Shield RP18 Column, 90Å, 1.6 μm, 3 mm × 100 mm
Catalog No. 502392270 Shop All Waters Corporation Products
Encompass_Preferred
CORTECS Shield RP18 columns offer complementary selectivity compared to C8 and C18 stationary phases. The embedded carbamate group in the bonded phase ligand provides alternative selectivity, particularly for phenolic compounds, when compared to straight-chain alkyl columns. The solid-core particle morphology results in high efficiency and lower backpressures compared to fully porous particles of similar size.
  • Offers complementary selectivity compared to C8 and C18 stationary phases.
  • Provides alternative selectivity, particularly for phenolic compounds, due to embedded carbamate group.
  • Delivers high efficiency with solid-core particle morphology.
  • Achieves lower backpressures compared to fully porous particles of similar size.
Catalog No. 50-239-2270 Supplier Waters Corp Supplier No. 176003935
CORTECS Shield RP18 columns offer complementary selectivity compared to C8 and C18 stationary phases. The embedded carbamate group in the bonded phase ligand provides alternative selectivity, particularly for phenolic compounds, when compared to straight-chain alkyl columns. The solid-core particle morphology results in high efficiency and lower backpressures compared to fully porous particles of similar size.
  • Offers complementary selectivity compared to C8 and C18 stationary phases.
  • Provides alternative selectivity, particularly for phenolic compounds, due to embedded carbamate group.
  • Delivers high efficiency with solid-core particle morphology.
  • Achieves lower backpressures compared to fully porous particles of similar size.