Learn More
Waters Corp CORTECS Shield RP18 Column, 90Å, 2.7 μm, 4.6 mm x 30 mm
Catalog No. 502392477 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
CORTECS Shield RP18 columns provide complementary selectivity to C8 and C18 stationary phases. The embedded carbamate group in the bonded phase ligand offers alternative selectivity, especially for phenolic compounds, compared to straight-chain alkyl columns. The solid-core particle morphology delivers high efficiency and lower backpressures when compared to fully porous particles of similar particle size.
  • Provides complimentary selectivity to C8 and C18 stationary phases.
  • Embedded carbamate group in the bonded phase ligand offers alternate selectivity, particularly for phenolic compounds, compared to straight chain alkyl columns.
  • Solid core particle morphology delivers high efficiency and lower backpressures when compared to fully porous particles of similar particle size.
Catalog No. 50-239-2477 Supplier Waters Corp Supplier No. 176003922
Add to Cart
Add to Cart
CORTECS Shield RP18 columns provide complementary selectivity to C8 and C18 stationary phases. The embedded carbamate group in the bonded phase ligand offers alternative selectivity, especially for phenolic compounds, compared to straight-chain alkyl columns. The solid-core particle morphology delivers high efficiency and lower backpressures when compared to fully porous particles of similar particle size.
  • Provides complimentary selectivity to C8 and C18 stationary phases.
  • Embedded carbamate group in the bonded phase ligand offers alternate selectivity, particularly for phenolic compounds, compared to straight chain alkyl columns.
  • Solid core particle morphology delivers high efficiency and lower backpressures when compared to fully porous particles of similar particle size.