Learn More
Waters Corp CORTECS Column, Shield RP18, 90Å, 2.7μm, 4.6 mm × 100 mm
Supplier:  Waters Corp 186008684
Encompass_Preferred
CORTECS Shield RP18 columns provide complementary selectivity to C8 and C18 stationary phases, using an embedded carbamate group for alternate selectivity, especially for phenolic compounds. Their solid core particle morphology delivers high efficiency and lower backpressures. This versatile product offers high-efficiency results, balanced retention of acids, bases, and neutrals, and excellent peak shape for basic analytes.
  • Delivers high-efficiency and versatile performance.
  • Offers balanced retention for various analytes across pH ranges.
  • Ensures excellent peak shape for basic analytes.
  • Resolves complex mixtures or maintains resolution while increasing throughput.
  • Solid-core particles improve performance by lowering van Deemter equation terms.
  • Increases sample throughput and reduces lab operation costs.
Catalog No. 50-239-0645