Learn More
Waters Corp CORTECS Shield RP18 Column, 90 Å, 1.6 μm, 3.0 x 50 mm
Supplier: Waters Corp 186008702
The CORTECS Shield RP18 Column is designed for high-performance liquid chromatography applications. It features a solid-core particle and provides high column efficiencies, resulting in sharper, narrower peaks for increased resolution, speed, and sensitivity.
- Shield RP18, solid-core particle
- USP class no. L1
- Ligand density: 3.2 μmol/m²
- Carbon load: 6.4%
- Endcapped: Yes
- pH range: 2–8
- Temperature limits: Low pH = 45 °C, high pH = 45 °C
- Surface area: 100 m²/g
- Performance standard: Neutrals QC Reference Material
- Application standard: Reversed-Phase QC Reference Material
- CORTECS UPLC Columns provide high efficiencies with low-dispersive UPLC instrumentation, producing sharper peaks for increased resolution, speed, and sensitivity.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.