Learn More
Waters Corp CORTECS UPLC Column, Shield RP18, 2.7 μm, 2.1 x 75 mm
Catalog No. 502392479 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The CORTECS Shield RP18 column is a UPLC column designed for the rapid separation of a wide array of compounds. Featuring 2.7 μm solid-core particle technology, it is suitable for use with HPLC and UHPLC systems.
  • Achieve high column efficiencies with low-dispersive UPLC instrumentation due to solid-core particle technology.
  • Produce sharper, narrower peaks compared to fully porous particles of similar size.
  • Increased resolution, speed, and sensitivity for enhanced performance.
  • Offers a variety of chemistries for flexible separation of various compounds.
Catalog No. 50-239-2479 Supplier Waters Corp Supplier No. 176003914
The CORTECS Shield RP18 column is a UPLC column designed for the rapid separation of a wide array of compounds. Featuring 2.7 μm solid-core particle technology, it is suitable for use with HPLC and UHPLC systems.
  • Achieve high column efficiencies with low-dispersive UPLC instrumentation due to solid-core particle technology.
  • Produce sharper, narrower peaks compared to fully porous particles of similar size.
  • Increased resolution, speed, and sensitivity for enhanced performance.
  • Offers a variety of chemistries for flexible separation of various compounds.