Learn More
Waters Corp CORTECS Column, 90Å, 2.7 μm, 3 mm X 150 mm
Catalog No. 502392800 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These Shield RP18 columns offer complementary selectivity to C8 and C18 stationary phases. The embedded carbamate group in the ligand provides alternate selectivity, especially for phenolic compounds. Solid core particle morphology ensures high efficiency and lower backpressures compared to fully porous particles.
  • Provides excellent peak shapes for basic analytes at low pH, offering balanced retention of bases, acids, and neutrals at low and mid-range pH.
  • Improves performance by reducing axial diffusion, ensuring more uniform packing, and enhancing heat transport.
  • Allows for increased sample throughput and helps lower laboratory costs by enabling simultaneous sample runs.
  • Can be used with a compatible guard cartridge to filter unwanted contaminants.
Catalog No. 50-239-2800 Supplier Waters Corp Supplier No. 186008675
These Shield RP18 columns offer complementary selectivity to C8 and C18 stationary phases. The embedded carbamate group in the ligand provides alternate selectivity, especially for phenolic compounds. Solid core particle morphology ensures high efficiency and lower backpressures compared to fully porous particles.
  • Provides excellent peak shapes for basic analytes at low pH, offering balanced retention of bases, acids, and neutrals at low and mid-range pH.
  • Improves performance by reducing axial diffusion, ensuring more uniform packing, and enhancing heat transport.
  • Allows for increased sample throughput and helps lower laboratory costs by enabling simultaneous sample runs.
  • Can be used with a compatible guard cartridge to filter unwanted contaminants.