Learn More
Waters Corp CORTECS Shield RP18 Column, 2.7 μm, 2.1 mm x 30 mm
Catalog No. 502391231 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
CORTECS Shield RP18 columns provide complementary selectivity to C8 and C18 stationary phases. The embedded carbamate group in the bonded phase ligand offers alternate selectivity, especially for phenolic compounds, compared to straight-chain alkyl columns. The solid core particle morphology provides high efficiency and lower backpressures when compared to fully porous particles of similar particle size.
  • Complementary selectivity to C8 and C18 stationary phases
  • Embedded carbamate group for alternate selectivity, especially for phenolic compounds
  • Solid core particle morphology for high efficiency and lower backpressures
Catalog No. 50-239-1231 Supplier Waters Corp Supplier No. 186008661
CORTECS Shield RP18 columns provide complementary selectivity to C8 and C18 stationary phases. The embedded carbamate group in the bonded phase ligand offers alternate selectivity, especially for phenolic compounds, compared to straight-chain alkyl columns. The solid core particle morphology provides high efficiency and lower backpressures when compared to fully porous particles of similar particle size.
  • Complementary selectivity to C8 and C18 stationary phases
  • Embedded carbamate group for alternate selectivity, especially for phenolic compounds
  • Solid core particle morphology for high efficiency and lower backpressures