Learn More
Waters Corp CORTECS Shield RP18 Column, 90Å, 2.7 μm, 2.1 mm, 150 mm
Catalog No. 502391303 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These columns offer complementary selectivity for phenolic compounds, high efficiency, and low backpressures due to solid core particle technology. Designed for reliable HPLC separations, they prevent unwanted surface interactions and ensure consistent batch results for easier method validation.
  • Complementary selectivity to C8 and C18 stationary phases.
  • Embedded carbamate group enhances separation of phenolic compounds.
  • Solid core particles provide high efficiency and lower backpressures.
  • Reliable performance from state-of-the-art packing and end-capping.
  • Supports flexible method development for complex separations.
  • Prevents unwanted surface-analyte interactions, aiding MS adoption.
  • Effective across varying conditions without compromising signal intensity or efficiency.
  • Consistent batch results simplify method validation and transfer.
Catalog No. 50-239-1303 Supplier Waters Corp Supplier No. 176003916
These columns offer complementary selectivity for phenolic compounds, high efficiency, and low backpressures due to solid core particle technology. Designed for reliable HPLC separations, they prevent unwanted surface interactions and ensure consistent batch results for easier method validation.
  • Complementary selectivity to C8 and C18 stationary phases.
  • Embedded carbamate group enhances separation of phenolic compounds.
  • Solid core particles provide high efficiency and lower backpressures.
  • Reliable performance from state-of-the-art packing and end-capping.
  • Supports flexible method development for complex separations.
  • Prevents unwanted surface-analyte interactions, aiding MS adoption.
  • Effective across varying conditions without compromising signal intensity or efficiency.
  • Consistent batch results simplify method validation and transfer.