Learn More
Waters Corp UPLC Shield RP18 Column, 90 Å, 2.7 μm, 3 × 150 mm
Catalog No. 502390651 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Waters CORTECS UPLC Shield RP18 Column leverages solid-core particle technology to deliver high-efficiency separations. Designed for increased resolution, speed, and sensitivity, this column provides sharper, narrower peaks compared to fully porous particles of similar size, making it ideal for demanding LC-MS applications requiring robust and reproducible results.
  • Solid-core particle technology for high column efficiencies
  • Endcapped Shield RP18 chemistry
  • USP Class No. L1 classification
  • Increased resolution, speed, and sensitivity
  • Sharper, narrower peaks for higher peak capacity
  • Excellent and reproducible LC-MS chromatograms
  • Robust and reliable performance
Catalog No. 50-239-0651 Supplier Waters Corp Supplier No. 176003936
The Waters CORTECS UPLC Shield RP18 Column leverages solid-core particle technology to deliver high-efficiency separations. Designed for increased resolution, speed, and sensitivity, this column provides sharper, narrower peaks compared to fully porous particles of similar size, making it ideal for demanding LC-MS applications requiring robust and reproducible results.
  • Solid-core particle technology for high column efficiencies
  • Endcapped Shield RP18 chemistry
  • USP Class No. L1 classification
  • Increased resolution, speed, and sensitivity
  • Sharper, narrower peaks for higher peak capacity
  • Excellent and reproducible LC-MS chromatograms
  • Robust and reliable performance