Learn More
Waters Corp CORTECS Shield RP18 Method Validation Kit, 90Å, 2.7μm, 3 mm × 100 mm
Catalog No. 502392111 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This method validation kit provides columns of three different batches, designed to offer complimentary selectivity to C8 and C18 stationary phases. The embedded carbamate group in the bonded phase ligand offers alternate selectivity, particularly for phenolic compounds. Its solid core particle morphology ensures high efficiency and lower backpressures compared to fully porous particles of similar size.
  • Provides columns of three different batches.
  • Offers complimentary selectivity to C8 and C18 stationary phases.
  • Embedded carbamate group in the bonded phase ligand provides alternate selectivity, especially for phenolic compounds compared to straight chain alkyl columns.
  • Solid core particle morphology offers high efficiency and lower backpressures when compared to fully porous particles of similar particle size.
Catalog No. 50-239-2111 Supplier Waters Corp Supplier No. 186008679
This method validation kit provides columns of three different batches, designed to offer complimentary selectivity to C8 and C18 stationary phases. The embedded carbamate group in the bonded phase ligand offers alternate selectivity, particularly for phenolic compounds. Its solid core particle morphology ensures high efficiency and lower backpressures compared to fully porous particles of similar size.
  • Provides columns of three different batches.
  • Offers complimentary selectivity to C8 and C18 stationary phases.
  • Embedded carbamate group in the bonded phase ligand provides alternate selectivity, especially for phenolic compounds compared to straight chain alkyl columns.
  • Solid core particle morphology offers high efficiency and lower backpressures when compared to fully porous particles of similar particle size.