Learn More
Waters Corp Method Validation Kit, CORTECS Shield RP18, 1.6 μm, 3 mm x 30 mm, 3 pk
Catalog No. 502392519 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This method validation kit includes columns from three different batches. The columns offer complementary selectivity to C8 and C18 stationary phases. The embedded carbamate group in the bonded phase ligand provides alternative selectivity, particularly for phenolic compounds, compared to straight chain alkyl columns. Its solid core particle morphology ensures high efficiency and lower backpressures compared to fully porous particles of similar size.
  • Complimentary selectivity to C8 and C18 stationary phases.
  • Embedded carbamate group in the bonded phase ligand for alternate selectivity, particularly for phenolic compounds.
  • Solid core particle morphology for high efficiency and lower backpressures.
Catalog No. 50-239-2519 Supplier Waters Corp Supplier No. 186008706
This method validation kit includes columns from three different batches. The columns offer complementary selectivity to C8 and C18 stationary phases. The embedded carbamate group in the bonded phase ligand provides alternative selectivity, particularly for phenolic compounds, compared to straight chain alkyl columns. Its solid core particle morphology ensures high efficiency and lower backpressures compared to fully porous particles of similar size.
  • Complimentary selectivity to C8 and C18 stationary phases.
  • Embedded carbamate group in the bonded phase ligand for alternate selectivity, particularly for phenolic compounds.
  • Solid core particle morphology for high efficiency and lower backpressures.