Learn More
Waters Corp CORTECS Method Validation Kit, 1.6 μm, 2.1 mm x 100 mm, 3/pk
Catalog No. 502392545 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
CORTECS Shield RP18 method validation kits provide columns of three different batches. These columns offer complementary selectivity to C8 and C18 stationary phases. The embedded carbamate group in the bonded phase ligand provides alternate selectivity, especially for phenolic compounds, compared to straight chain alkyl columns. The solid core particle morphology offers high efficiency and lower backpressures when compared to fully porous particles of similar particle size.
  • Provides complimentary selectivity to C8 and C18 stationary phases.
  • Offers alternate selectivity, especially for phenolic compounds.
  • Solid core particle morphology offers high efficiency.
  • Solid core particle morphology offers lower backpressures.
Catalog No. 50-239-2545 Supplier Waters Corp Supplier No. 186008699
CORTECS Shield RP18 method validation kits provide columns of three different batches. These columns offer complementary selectivity to C8 and C18 stationary phases. The embedded carbamate group in the bonded phase ligand provides alternate selectivity, especially for phenolic compounds, compared to straight chain alkyl columns. The solid core particle morphology offers high efficiency and lower backpressures when compared to fully porous particles of similar particle size.
  • Provides complimentary selectivity to C8 and C18 stationary phases.
  • Offers alternate selectivity, especially for phenolic compounds.
  • Solid core particle morphology offers high efficiency.
  • Solid core particle morphology offers lower backpressures.