Learn More
Waters Corp Method Validation Kit, CORTECS Shield RP18, 1.6 μm, 3.0 × 100 mm
Catalog No. 502391481 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This Method Validation Kit is designed for CORTECS Shield RP18 columns, featuring a particle size of 1.6 μm and dimensions of 3.0 × 100 mm. Each kit provides three columns from different batches of material for comprehensive method validation.
  • Kit contains three columns from three batches of material.
  • Designed for CORTECS Shield RP18 columns.
  • Features 1.6 μm particle size.
  • Column dimensions are 3.0 × 100 mm.
Catalog No. 50-239-1481 Supplier Waters Corp Supplier No. 186008709
This Method Validation Kit is designed for CORTECS Shield RP18 columns, featuring a particle size of 1.6 μm and dimensions of 3.0 × 100 mm. Each kit provides three columns from different batches of material for comprehensive method validation.
  • Kit contains three columns from three batches of material.
  • Designed for CORTECS Shield RP18 columns.
  • Features 1.6 μm particle size.
  • Column dimensions are 3.0 × 100 mm.