Learn More
Waters Corp UPLC Column, Shield RP18, 1.6 μm, 2.1 x 30 mm
Catalog No. 502391909 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The CORTECS UPLC Column, Shield RP18, is provided as part of a Method Validation Kit. It features a 1.6 μm particle size and 2.1 x 30 mm dimensions, designed for high-speed applications and efficient separation of various compounds.
  • Kit includes three columns from three material batches.
  • 90 Å pore size.
  • 1.6 μm particle size.
  • 2.1 x 30 mm column dimensions.
  • 3.2 μmol/m² ligand density.
  • 6.4% carbon load.
  • Endcapped stationary phase.
  • pH range of 2–8.
  • Temperature limits: 45 °C for low/high pH.
  • 100 m²/g surface area.
  • High column efficiencies with low-dispersive UPLC instrumentation.
  • Produces sharper, narrower peaks than fully porous particles.
  • Ideal for increased resolution, speed, and sensitivity.
Catalog No. 50-239-1909 Supplier Waters Corp Supplier No. 186008696
The CORTECS UPLC Column, Shield RP18, is provided as part of a Method Validation Kit. It features a 1.6 μm particle size and 2.1 x 30 mm dimensions, designed for high-speed applications and efficient separation of various compounds.
  • Kit includes three columns from three material batches.
  • 90 Å pore size.
  • 1.6 μm particle size.
  • 2.1 x 30 mm column dimensions.
  • 3.2 μmol/m² ligand density.
  • 6.4% carbon load.
  • Endcapped stationary phase.
  • pH range of 2–8.
  • Temperature limits: 45 °C for low/high pH.
  • 100 m²/g surface area.
  • High column efficiencies with low-dispersive UPLC instrumentation.
  • Produces sharper, narrower peaks than fully porous particles.
  • Ideal for increased resolution, speed, and sensitivity.