Learn More
Waters Corp UPLC Column Kit, Shield RP18, 4.6 × 100 mm
Catalog No. 502392163 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This Method Validation Kit includes three columns for evaluating the quality, reliability, and consistency of chromatographic methods. Each kit contains three columns derived from different base particles.
  • Includes three columns with 4.6 mm × 100 mm dimensions.
  • Features 2.7 μm particle size.
  • Designed with 90 Å pore size.
  • Each kit contains three columns from three batches of material.
Catalog No. 50-239-2163 Supplier Waters Corp Supplier No. 186008689
This Method Validation Kit includes three columns for evaluating the quality, reliability, and consistency of chromatographic methods. Each kit contains three columns derived from different base particles.
  • Includes three columns with 4.6 mm × 100 mm dimensions.
  • Features 2.7 μm particle size.
  • Designed with 90 Å pore size.
  • Each kit contains three columns from three batches of material.