Learn More
Waters Corp CORTECS Shield RP18 Kit, 2.7 μm, 4.6 mm × 50 mm
Catalog No. 502392324 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
CORTECS UPLC Columns Method Validation Kits are designed to evaluate the quality, reliability, and consistency of analytical methods. These kits include three batches of chromatographic media derived from different base particles, with each kit containing three columns from three distinct batches of material.
  • Shield RP18 column chemistry.
  • Particle size of 2.7 μm.
  • Column dimensions of 4.6 mm × 50 mm.
  • Designed to evaluate the quality, reliability, and consistency of analytical methods.
  • Includes three batches of chromatographic media.
  • Contains three columns from three distinct batches of material.
Catalog No. 50-239-2324 Supplier Waters Corp Supplier No. 186008687
CORTECS UPLC Columns Method Validation Kits are designed to evaluate the quality, reliability, and consistency of analytical methods. These kits include three batches of chromatographic media derived from different base particles, with each kit containing three columns from three distinct batches of material.
  • Shield RP18 column chemistry.
  • Particle size of 2.7 μm.
  • Column dimensions of 4.6 mm × 50 mm.
  • Designed to evaluate the quality, reliability, and consistency of analytical methods.
  • Includes three batches of chromatographic media.
  • Contains three columns from three distinct batches of material.