Learn More
Waters Corp Method Validation Kit, CORTECS Shield RP18, 90Å, 1.6 μm
Catalog No. 502391055 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These method validation kits provide columns of three different batches. The columns offer complementary selectivity to C8 and C18 stationary phases. The embedded carbamate group in the bonded phase ligand provides alternate selectivity, especially for phenolic compounds, compared to straight chain alkyl columns. The solid core particle morphology offers high efficiency and lower backpressures when compared to fully porous particles of similar particle size.
  • Provides columns of three different batches.
  • Offers complementary selectivity to C8 and C18 stationary phases.
  • Embedded carbamate group in the bonded phase ligand provides alternate selectivity, especially for phenolic compounds.
  • Solid core particle morphology offers high efficiency.
  • Solid core particle morphology offers lower backpressures when compared to fully porous particles of similar particle size.
Catalog No. 50-239-1055 Supplier Waters Corp Supplier No. 186008708
These method validation kits provide columns of three different batches. The columns offer complementary selectivity to C8 and C18 stationary phases. The embedded carbamate group in the bonded phase ligand provides alternate selectivity, especially for phenolic compounds, compared to straight chain alkyl columns. The solid core particle morphology offers high efficiency and lower backpressures when compared to fully porous particles of similar particle size.
  • Provides columns of three different batches.
  • Offers complementary selectivity to C8 and C18 stationary phases.
  • Embedded carbamate group in the bonded phase ligand provides alternate selectivity, especially for phenolic compounds.
  • Solid core particle morphology offers high efficiency.
  • Solid core particle morphology offers lower backpressures when compared to fully porous particles of similar particle size.