Learn More
Waters Corp CORTECS VanGuard Cartridge, 90Å, 2.7 μm, 2.1 mm x 5 mm
Catalog No. 502391556 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
CORTECS Shield RP18 VanGuard Cartridges are designed to extend analytical column lifetime and performance by removing particulate contamination from the mobile phase stream. This cartridge is optimized to protect all 2.1 mm and 3.0 mm I.D. CORTECS Shield RP18 analytical columns containing 2.7 μm sorbent particles.
  • Extends analytical column lifetime and performance.
  • Improves separation efficiency for sharper peaks and better resolution.
  • Reduces analysis time, increasing laboratory productivity.
  • Ensures consistent performance and enhanced reproducibility.
  • Constructed from high-quality materials for durability.
  • Backed by a comprehensive warranty.
  • Offers precise and accurate analysis capabilities.
  • Compatible with a wide range of mobile phases.
  • Convenient 3-pack for uninterrupted productivity.
Catalog No. 50-239-1556 Supplier Waters Corp Supplier No. 186008712
CORTECS Shield RP18 VanGuard Cartridges are designed to extend analytical column lifetime and performance by removing particulate contamination from the mobile phase stream. This cartridge is optimized to protect all 2.1 mm and 3.0 mm I.D. CORTECS Shield RP18 analytical columns containing 2.7 μm sorbent particles.
  • Extends analytical column lifetime and performance.
  • Improves separation efficiency for sharper peaks and better resolution.
  • Reduces analysis time, increasing laboratory productivity.
  • Ensures consistent performance and enhanced reproducibility.
  • Constructed from high-quality materials for durability.
  • Backed by a comprehensive warranty.
  • Offers precise and accurate analysis capabilities.
  • Compatible with a wide range of mobile phases.
  • Convenient 3-pack for uninterrupted productivity.