Learn More
Waters Corp Column, CORTECS T3, 1.6 μm, 2.1 mm x 150 mm
Catalog No. 502392216 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
CORTECS T3 columns are compatible with 100% aqueous mobile phases and provide excellent retention for polar and non-polar analytes. The particle morphology offers high efficiency and lower backpressure compared to fully porous particles of similar particle size. These columns provide ultra-low column MS bleed, long lifetimes, and excellent peak shape, increasing throughput and speed of analysis without sacrificing separation performance.
  • Increase throughput and achieve higher efficiency by raising the method’s flow rate.
  • Help decrease analysis and lab equipment costs by running more samples and obtaining results faster.
  • Manufactured in an ISO 9001 certified facility using pure reagents and strict controls.
Catalog No. 50-239-2216 Supplier Waters Corp Supplier No. 176003895
CORTECS T3 columns are compatible with 100% aqueous mobile phases and provide excellent retention for polar and non-polar analytes. The particle morphology offers high efficiency and lower backpressure compared to fully porous particles of similar particle size. These columns provide ultra-low column MS bleed, long lifetimes, and excellent peak shape, increasing throughput and speed of analysis without sacrificing separation performance.
  • Increase throughput and achieve higher efficiency by raising the method’s flow rate.
  • Help decrease analysis and lab equipment costs by running more samples and obtaining results faster.
  • Manufactured in an ISO 9001 certified facility using pure reagents and strict controls.