Learn More
Waters Corp Column, C18, 2.1 mm × 75 mm, 1.6 μm, 120 Å
Catalog No. 502391715 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
CORTECS T3 columns are compatible with 100% aqueous mobile phases and offer excellent retention for both polar and non-polar analytes. The particle morphology provides high efficiency and lower backpressure compared to fully porous particles of similar size. It also ensures ultra-low column MS bleed, long lifetimes, and excellent peak shape.
  • Compatible with 100% aqueous mobile phases.
  • Provides excellent retention for polar and non-polar analytes.
  • Offers high efficiency.
  • Results in lower backpressure compared to fully porous particles of similar size.
  • Ensures ultra-low column MS bleed.
  • Provides long lifetimes.
  • Delivers excellent peak shape.
Catalog No. 50-239-1715 Supplier Waters Corp Supplier No. 186008498
CORTECS T3 columns are compatible with 100% aqueous mobile phases and offer excellent retention for both polar and non-polar analytes. The particle morphology provides high efficiency and lower backpressure compared to fully porous particles of similar size. It also ensures ultra-low column MS bleed, long lifetimes, and excellent peak shape.
  • Compatible with 100% aqueous mobile phases.
  • Provides excellent retention for polar and non-polar analytes.
  • Offers high efficiency.
  • Results in lower backpressure compared to fully porous particles of similar size.
  • Ensures ultra-low column MS bleed.
  • Provides long lifetimes.
  • Delivers excellent peak shape.