Learn More
Waters Corp CORTECS Column, C18, 1.6 μm, 120 Å, 2.1 mm x 100 mm
Catalog No. 502391818 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These columns are compatible with 100% aqueous mobile phases and provide excellent retention for polar and non-polar analytes. The particle morphology offers high efficiency and lower backpressure compared to fully porous particles of similar size. They provide ultra-low column MS bleed, long lifetimes, and excellent peak shape.
  • C18 chemistry for reversed phase separation.
  • Silica particle substrate; spherical shape; low silanol activity.
  • PH range of 2 to 8.
  • Maximum temperature limit of 45°C.
  • Maximum pressure of 18000 psi (1240 bar).
  • Endcapped design with T3 bonding technology.
  • 1.6 μm particle size; 120 Å pore size.
  • Compatible with UPLC and UHPLC systems.
  • Solid core particle technology; L1 USP classification.
  • 2.1 mm inner diameter; 100 mm length.
  • 4.7% carbon load.
  • Equipped with eCord.
  • Available in a 3 pack.
Catalog No. 50-239-1818 Supplier Waters Corp Supplier No. 176003894
These columns are compatible with 100% aqueous mobile phases and provide excellent retention for polar and non-polar analytes. The particle morphology offers high efficiency and lower backpressure compared to fully porous particles of similar size. They provide ultra-low column MS bleed, long lifetimes, and excellent peak shape.
  • C18 chemistry for reversed phase separation.
  • Silica particle substrate; spherical shape; low silanol activity.
  • PH range of 2 to 8.
  • Maximum temperature limit of 45°C.
  • Maximum pressure of 18000 psi (1240 bar).
  • Endcapped design with T3 bonding technology.
  • 1.6 μm particle size; 120 Å pore size.
  • Compatible with UPLC and UHPLC systems.
  • Solid core particle technology; L1 USP classification.
  • 2.1 mm inner diameter; 100 mm length.
  • 4.7% carbon load.
  • Equipped with eCord.
  • Available in a 3 pack.