Learn More
Waters Corp Column, C18, 2.7 μm, 120 Å, 3 mm x 75 mm
Catalog No. 502391647 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
CORTECS T3 columns are designed for high-performance liquid chromatography, offering exceptional retention for a wide range of analytes. Featuring advanced particle morphology, these columns deliver high efficiency and lower backpressure, making them suitable for demanding analytical applications.
  • Compatible with 100% aqueous mobile phases.
  • Provides excellent retention for both polar and non-polar analytes.
  • Offers high efficiency and lower backpressure compared to fully porous particles of similar particle size.
  • Delivers ultra-low column MS bleed.
  • Ensures long lifetimes and excellent peak shape.
Catalog No. 50-239-1647 Supplier Waters Corp Supplier No. 176003883
CORTECS T3 columns are designed for high-performance liquid chromatography, offering exceptional retention for a wide range of analytes. Featuring advanced particle morphology, these columns deliver high efficiency and lower backpressure, making them suitable for demanding analytical applications.
  • Compatible with 100% aqueous mobile phases.
  • Provides excellent retention for both polar and non-polar analytes.
  • Offers high efficiency and lower backpressure compared to fully porous particles of similar particle size.
  • Delivers ultra-low column MS bleed.
  • Ensures long lifetimes and excellent peak shape.